Anti-ADCK4 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8070R
10476-116EA 796.88 CAD
10476-116
Anti-ADCK4 Rabbit Polyclonal Antibody
Antibodies
ADCK4, AarF domain containing kinase 5, is a member of group i collagen (fibrillar forming collagen). The function of this protein is not yet clear. It is not known if it has protein kinase activity and what type of substrate it would phosphorylate (Ser, Thr or Tyr). There are 2 named isoforms produced by alternative splicing.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADCK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR