Anti-CCDC124 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8083R
10476-396EA 796.88 CAD
10476-396
Anti-CCDC124 Rabbit Polyclonal Antibody
Antibodies
CCDC124

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCDC124
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR