Anti-CCDC144A Rabbit Polyclonal Antibody

BS-8106R
10476-628EA 407.07 CAD
10476-628
Anti-CCDC144A Rabbit Polyclonal Antibody
CCDC144A

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCDC144A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR