Anti-CCDC144NL Rabbit Polyclonal Antibody

Supplier: Bioss
BS-8115R
10477-148EA 677.18 CAD
10477-148
Anti-CCDC144NL Rabbit Polyclonal Antibody
CCDC144NL

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCDC144NL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR