Anti-CCDC22 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8124R
10477-108EA 796.88 CAD
10477-108
Anti-CCDC22 Rabbit Polyclonal Antibody
Antibodies
This gene encodes a protein containing a coiled-coil domain. The mouse orthologous protein has been shown to bind copines, which are calcium-dependent, membrane-binding proteins that may function in calcium signaling. Localization of the orthologous rat protein suggests that it may play a role in neuronal injury response. This human gene has been identified as a novel candidate gene for syndromic X-linked intellectual disability. [provided by RefSeq, Aug 2011].

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCDC22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR