Anti-CCDC38 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8130R
10477-508EA 796.88 CAD
10477-508
Anti-CCDC38 Rabbit Polyclonal Antibody
Antibodies
The coiled-coil domain is a structural motif found in proteins that are involved in a diverse array of biological functions such as the regulation of gene expression, cell division, membrane fusion and drug extrusion and delivery. CCDC38 (coiled-coil domain containing 38) is a 563 amino acid protein encoded by a gene that maps to human chromosome 12q23.1. Encoding over 1,100 genes, chromosome 12 comprises approximately 4.5% of the human genome. Chromosome 12 is associated with a variety of diseases and afflictions, including hypochondrogenesis, achondrogenesis, Kniest dysplasia, Noonan syndrome and trisomy 12p, which causes facial developmental defects and seizure disorders.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCDC38
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR