Anti-BTBD1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8158R
10478-102EA 796.88 CAD
10478-102
Anti-BTBD1 Rabbit Polyclonal Antibody
Antibodies
BTBD1 contains a proline-rich region and a BTB/POZ domain which are typically involved in protein-protein interactions. The C-terminus of BTBD1 binds topoisomerase I and BTBD1 also interacts with TRIM5 isoform Delta. BTBD1 localizes to cytoplasmic bodies and is essential for myogenic differentiation.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: BTBD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR