Anti-TCAB1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8163R
10478-270EA 796.88 CAD
10478-270
Anti-TCAB1 Rabbit Polyclonal Antibody
Antibodies
WDR79 contains six WD (tryptophan-aspartate) repeat domains found in a number of proteins that function as adaptor molecules in signal transduction and cytoskeletal organization. The WD repeat is defined by four or more repeating units of a conserved core of approximately 40 amino acids ending with tryptophan-aspartic acid (WD). WD repeats may serve as sites of protein-protein interaction for adaptor proteins and facilitate multiprotein complex formation. The function of the WDR79 protein has not been characterized, however significant and consistent single nucleotide polymorphisms in the WDR79 gene have been found to be associated with ER negative breast cancer.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TCAB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR