Anti-CHCHD6 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8167R
10478-166EA 796.88 CAD
10478-166
Anti-CHCHD6 Rabbit Polyclonal Antibody
Antibodies
CHCHD6

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CHCHD6
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR