Anti-AVSD2 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8184R
10478-734EA 796.62 CAD
10478-734
Anti-AVSD2 Rabbit Polyclonal Antibody
Antibodies
CRELD1 is a protein composed of Epidermal growth factor-like repeats, which represent a group of cysteine-rich domains that mediate interactions between proteins of diverse function. Such domains are found in proteins that are either completely secreted or have transmembrane regions that link the protein to the cell surface.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: AVSD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR