Anti-CRELD2 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8185R
10478-758EA 796.88 CAD
10478-758
Anti-CRELD2 Rabbit Polyclonal Antibody
Antibodies
May regulate transport of alpha4-beta2 neuronal acetylcholine receptor.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRELD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR