Anti-CSMD3 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8190R
10478-880EA 796.88 CAD
10478-880
Anti-CSMD3 Rabbit Polyclonal Antibody
Antibodies
CSMD3 belongs to the CSMD family and contains fourteen CUB domains and twenty eight Sushi (CCP/SCR) domains. It is weakly expressed in most tissues, except in brain; expressed at intermediate level in brain, including cerebellum, substantia nigra, thalamus, spinal cord, hippocampus and fetal brain. Also expressed in testis. There are five named isoforms.

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CSMD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR