Anti-CTAGE4 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8193R
10479-008EA 796.88 CAD
10479-008
Anti-CTAGE4 Rabbit Polyclonal Antibody
Antibodies
Tumor-associated antigen.Tissue specificity:Expressed in testis, placenta and skin. Expressed at lower level in mammary gland and stomach.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CTAGE4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR