Anti-NXPE4 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8196R
10479-080EA 796.62 CAD
10479-080
Anti-NXPE4 Rabbit Polyclonal Antibody
Antibodies
FAM55D

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: NXPE4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR