Anti-FAM50A Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8208R
10479-218EA 796.88 CAD
10479-218
Anti-FAM50A Rabbit Polyclonal Antibody
Antibodies
FAM50A (Family with sequence similarity 50, member A) is a basic protein containing a nuclear localization signal. Its specifc function is unknown. It may be a DNA-binding protein or transcriptional factor.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: FAM50A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR