Anti-FAM21C Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8214R
10479-378EA 796.88 CAD
10479-378
Anti-FAM21C Rabbit Polyclonal Antibody
Antibodies
Playing a essential role in the fission of tubules that serve as transport intermediates during endosome sorting, the WASH complex is present at the surface of endosomes and functions to recruit and activate the Arp2/3 complex for induction of actin polymerization. FAM21, Putative WASH complex subunit FAM21, is a component of the WASH complex. Having undergone evolutionary duplication, four highly homologous family members exist including FAM21A, FAM21B, FAM21C and FAM21D. FAM21 links the WASH complex to endosomes and is required for WASM-dependent retromer-mediated sorting. Also, by directly interacting with CapZ, FAM21 inhibits its anti-capping activity, thereby regulating actin dynamics.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: FAM21C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR