Anti-CUTA Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8248R
10479-948EA 796.61 CAD
10479-948
Anti-CUTA Rabbit Polyclonal Antibody
Antibodies
May forms part of a complex of membrane proteins attached to acetylcholinesterase (AChE).Tissue specificity:Ubiquitous. Widely expressed in brain.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CUTA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR