Anti-GIMAP6 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8272R
10480-800EA 796.62 CAD
10480-800
Anti-GIMAP6 Rabbit Polyclonal Antibody
Antibodies
Highly expressed in spleen, lymph nodes, lung and placenta. Expressed at moderate level in thymus, kidney, heart and digestive tract. Weakly expressed in other lymphoid tissues.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: GIMAP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR