Anti-DNAJC7 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8280R
10480-736EA 796.88 CAD
10480-736
Anti-DNAJC7 Rabbit Polyclonal Antibody
Antibodies
The DnaJ family is one of the largest of all the chaperone families and has evolved with diverse cellular localization and functions. The presence of the J domain defines a protein as a member of the DnaJ family. DnaJ heat shock induced proteins are from the bacterium Escherichia coli and are under the control of the htpR regulatory protein. The DnaJ proteins play a critical role in the HSP 70 chaperone machine by interacting with HSP 70 to stimulate ATP hydrolysis. The proteins contain cysteine rich regions that are composed of zinc fingers that form a peptide-binding domain responsible for the chaperone function. DnaJ proteins are important mediators of proteolysis and are in-volved in the regulation of protein degradation, exocytsis and endocytosis. DnaJC7 (DnaJ homolog subfamily C member 7), also known as TPR2, TTC2 or DANJC7, is ubiquitously expressed, with highest expression in testis, liver, heart and brain.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: DNAJC7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR