Anti-TMPRSS6 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8296R
10481-320EA 796.86 CAD
10481-320
Anti-TMPRSS6 Rabbit Polyclonal Antibody
Antibodies
Serine protease which hydrolyzes a range of proteins including type I collagen, fibronectin and fibrinogen. Can also activate urokinase-type plasminogen activator with low efficiency. May play a specialized role in matrix remodeling processes in liver. Required to sense iron deficiency. Overexpression suppresses activation of the HAMP promoter.Involvement in disease:Defects in TMPRSS6 are the cause of iron-refractory iron deficiency anemia (IRIDA); also known as hypochromic microcytic anemia with defect in iron metabolism or hereditary iron-handling disorder or pseudo-iron-deficiency anemia. Key features include congenital hypochromic microcytic anemia, very low mean corpuscular erythrocyte volume, low transferrin saturation, abnormal iron absorption characterized by no hematologic improvement following treatment with oral iron, and abnormal iron utilization characterized by a sluggish, incomplete response to parenteral iron.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TMPRSS6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR