Anti-FAM46D Rabbit Polyclonal Antibody

BS-8313R
10481-474EA 397.94 CAD
10481-474
Anti-FAM46D Rabbit Polyclonal Antibody
Antibodies against the protein encoded by this gene were found only in plasma from cancer patients. While it may be a target for immunotherapy, the function of this protein is unknown.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: FAM46D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR