Anti-HDHD3 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8318R
10481-778EA 796.88 CAD
10481-778
Anti-HDHD3 Rabbit Polyclonal Antibody
Antibodies
HDHD3 (haloacid dehalogenase-like hydrolase domain containing 3) is a 251 amino acid protein that belongs to the HAD-like hydrolase superfamily, which contains a group of hydrolase enzymes that differ from the å/∫ hydrolase family based on structure. This family of hydrolase enzymes includes L-2-haloacid dehalogenase, epoxide hydrolases and phosphatases. HDHD3 is encoded by a gene located on human chromosome 9q32. Chromosome 9 consists of about 145 million bases, 4% of the human genome and encodes nearly 900 genes. Thought to play a role in gender determination, deletion of the distal portion of 9p can lead to development of male to female sex reversal, the phenotype of a female with a male X,Y genotype.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: HDHD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR