Anti-PANK4 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8340R
10482-266EA 796.88 CAD
10482-266
Anti-PANK4 Rabbit Polyclonal Antibody
Antibodies
PANK4 belongs to the pantothenate kinase family. Pantothenate kinase is a key regulatory enzyme in the physiological regulation of the intracellular coenzyme A (CoA) concentration. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by CoA. This family member is most abundant in muscle but is expressed in all tissues.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: PANK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR