Anti-FGFR1OP2 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8346R
10482-066EA 796.88 CAD
10482-066
Anti-FGFR1OP2 Rabbit Polyclonal Antibody
Antibodies
FGFR1OP2 belongs to the SIKE family. The FGFR1OP2 (FGFR1 oncogene partner 2) gene was identified through its involvement in a fusion with the FGFR1 gene. FGFR1OP2 may be involved in the wound healing pathway. It is expressed in bone marrow, spleen and thymus. A chromosomal aberration involving FGFR1OP2 may be a cause of stem cell myeloproliferative disorder (MPD). Insertion ins(12;8)(p11;p11p22) with FGFR1. MPD is characterized by myeloid hyperplasia, eosinophilia and T cell or B cell lymphoblastic lymphoma. In general it progresses to acute myeloid leukemia. The fusion protein FGFR1OP2-FGFR1 may exhibit constitutive kinase activity and be responsible for the transforming activity.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: FGFR1OP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR