Anti-UBE2G2 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8349R
10482-474EA 796.88 CAD
10482-474
Anti-UBE2G2 Rabbit Polyclonal Antibody
Antibodies
UBE2G2 (Ubiquitin-conjugating enzyme E2 G2), also known as UBC7, is a 165 amino acid protein involved in ubiquitin-mediated protein degradation. Ubiquitination is an important mechanism through which three classes of enzymes act in concert to target short-lived or abnormal proteins for destruction. The three classes of enzymes involved in ubiquitination are the ubiquitin-activating enzymes (E1s), the ubiquitin-conjugating enzymes (E2s) and the ubiquitin-protein ligases (E3s). UBE2G2 is an E2 ubiquitin-conjugating enzyme that acts to catalyze the covalent attachment of ubiquitins to various proteins. Expressed throughout the body, UBE2G2 shares 100% sequence identity with its mouse counterpart and is thought to be involved in endoplasmic reticulum-associated degradation (ERAD). Two isoforms of UBE2G2 exist due to alternative splicing events.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: UBE2G2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR