Anti-LRRC2 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8360R
10482-522EA 796.88 CAD
10482-522
Anti-LRRC2 Rabbit Polyclonal Antibody
Antibodies
The leucine-rich repeat-containing protein 2 (LRRC2) is a 371 amino acid protein that contains 9 LRR repeats. The gene encoding LRRC2 maps to chromosome 3, which encodes over 1,100 genes. Particular regions of the chromosome 3 short arm are deleted in many types of cancer cells. Marfan Syndrome, porphyria, von Hippel-Lindau syndrome, osteogenesis imperfecta and Charcot-Marie-Tooth Disease are a few of the numerous genetic diseases associated with chromosome 3.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: LRRC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR