Anti-CBLL1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8386R
10483-050EA 796.89 CAD
10483-050
Anti-CBLL1 Rabbit Polyclonal Antibody
Antibodies
CBLL1, also known as HAKAI (meaning ‘destruction’ in Japanese), or RNF188 (RING finger protein 188), is a 491 amino acid protein that contains one C2H2-type zinc finger and one RING-type zinc finger. CBLL1 is believed to function as an E3 ubiquitin-protein ligase that accepts a ubiquitin residue from an E2 ubiquitin-conjugating enzyme and immediately transfers that residue to a protein that is targeted for degradation. More specifically, upon activation of c-Src, CBLL1 interacts with and ubiquitinates tyrosine-phosphorylated E-cadherin, thereby targeting the E-cadherin complex for endocytosis and disrupting epithelial cell-cell contacts. Via its role as an E-cadherin regulator, CBLL1 participates in cell adhesion and may also be involved in the regulation of epithelial-mesenchymal transitions.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CBLL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR