Anti-BTBD10 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8398R
10483-642EA 796.88 CAD
10483-642
Anti-BTBD10 Rabbit Polyclonal Antibody
Antibodies
The BTB (broad-complex, Tramtrack and Bric a brac) domain, also known as the POZ (Poxvirus and zinc finger) domain, is an N-terminal homodimerization domain that contains multiple copies of kelch repeats and/or C2H2-type zinc fingers. Proteins that contain BTB domains are thought to be involved in transcriptional regulation via control of chromatin structure and function. BTBD10 (BTB (POZ) domain containing 10), also known as GMRP1, is a ubiquitously expressed nuclear protein found at highest levels in adult testis, brain and small intestine and weakly expressed in colon, lung, liver, kidney, spleen,pancreas, thymus, prostate, heart and ovary. Down-regulated in glioma, BTBD10 binds PP2A (protein phosphatase 2A) to inhibit dephosphorylation of Akts and is suggested to be a suppressor of cell death as well as an enhancer of cell growth. BTBD10 contains one BTB (POZ) domain and is encoded by a gene mapping to human chromosome 11p15.2.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: BTBD10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR