Anti-GCLM Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8402R
10483-714EA 796.88 CAD
10483-714
Anti-GCLM Rabbit Polyclonal Antibody
Antibodies
The GCLC gene consists of 16 exons and encodes the 636 amino acid protein g-GCSc (g-glutamylcysteine synthetase heavy subunit), also designated g-L-glutamate-L-cysteine ligase catalytic subunit (GLCLC). g-GCSc is expressed in hemocytes, brain, liver and kidney. g-GCSc associates with a regulatory or modifier subunit, g-GCSm (g-glutamylcysteine synthetase light subunit), to form a heterodimer, g-GCS. g-GCS is the first enzyme involved and the rate determining step in glutathione biosynthesis. Oxidants, cadium and methyl mercury upregulate the transcription of g-GCS. H2O2 regulation depends on the Yap1 protein and the presence of glutamate, glutamine and lysine. Cadium regulates transcription through proteins Met-4, Met-31 and Met-32. Cbf1, a DNA binding protein, inhibits transcription of g-GCS. Chemopreventive compounds cause increased levels of g-GCSc in kidney tissues, which may protect against chemically induced carcinogenesis. A His370Leu amino acid change in g-GCSc causes deficiencies in activity which are responsible for hemolytic anemia and low red blood cell glutathione levels.Defects in GCLC are the cause of hemolytic anemia.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: GCLC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR