Anti-GLB1L3 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8409R
10483-858EA 796.62 CAD
10483-858
Anti-GLB1L3 Rabbit Polyclonal Antibody
Antibodies
GLB1L3 is a 653 amino acid protein belonging to the glycosyl hydrolase 35 family. GLB1L3 exists as three alternatively spliced isoforms and is encoded by a gene that maps to human chromosome 11q25. With approximately 135 million base pairs and 1,400 genes, chromosome 11 makes up around 4% of human genomic DNA and is considered a gene and disease association dense chromosome. The chromosome 11 encoded Atm gene is important for regulation of cell cycle arrest and apoptosis following double strand DNA breaks. Atm mutation leads to the disorder known as ataxia-telangiectasia. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are also associated with defects in chromosome 11.

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: GLB1L3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR