Anti-GAL12 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8413R
10483-954EA 796.86 CAD
10483-954
Anti-GAL12 Rabbit Polyclonal Antibody
Antibodies
Galectin-12, also designated galectin-related inhibitor of proliferation 1 in mouse, is a 314 amino acid protein encoded by the human gene LGALS12. Galectin-12 is a member of the galectin family consisting of b-galactoside-binding proteins with conserved carbohydrate recognition domains. Galectin-12 binds lactose and may participate in the apoptosis of adipocytes. This protein is preferentially expressed in peripheral blood leukocytes and adipocytes. Galectin-12 is induced by cell cycle block at the G1 phase and causes G1 arrest when overexpressed. The galectin-12 gene is expressed in mouse preadipo-cytes and is upregulated when preadipocytes undergo cell cycle arrest, concomitant with acquisition of the competence to undergo differentiation in response to adipogenic hormone stimulation. Galectin-12 is an adipocyte-expressed protein which is downregulated by various insulin resistance-inducing hormones. As a result, galectin-12 may play a role in the pathogenesis of insulin resistance.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: Galectin 12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR