Anti-HEATR7B2 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8415R
10484-058EA 796.88 CAD
10484-058
Anti-HEATR7B2 Rabbit Polyclonal Antibody
Antibodies
HEAT repeats form rod-like helical structures that are involved in intracellular transport. HEATR7B2 (HEAT repeat-containing protein 7B2) is a 1,585 amino acid protein that contains fifteen HEAT repeats. Existing as two alternatively spliced isoforms, the gene encoding HEATR7B2 maps to human chromosome 5p13.1 and mouse chromosome 15 A1. Chromosome 5 contains 181 million base pairs and comprises nearly 6% of the human genome. Cockayne syndrome is associated with chromosome 5 through the ERCC8 gene and familial adenomatous polyposis via the adenomatous polyposis coli (APC) tumor suppressor gene. Treacher Collins syndrome is also chromosome 5-associated and is caused by insertions or deletions within the TCOF1 gene. Deletion of the p arm of chromosome 5 leads to Cri du chat syndrome, while deletion of the q arm or of chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: HEATR7B2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR