Anti-FNDC1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8460R
10485-100EA 796.88 CAD
10485-100
Anti-FNDC1 Rabbit Polyclonal Antibody
Antibodies
FNDC1, also known as MEL4B3, Ags8, Expressed in synovial lining protein and Activation-associated cDNA protein, is a 1888 amino acid secreted protein that contains five fibronectin type-III domains. FNDC1 is moderately expressed in skeletal muscle, pancreas, heart, kidney, spinal cord, ovary and lung. Expression of FNDC1 is induced in response to hypoxia in ventricular cardiomyocytes. Since FNDC1 interacts with G∫ and G©, it is likely that FNDC1 is an activator for G-protein signaling. Though normally absent in healthy skin, FNDC1 expression is induced by TGF-beta signaling in skin tumors and psoriasis. There are two isoforms of FNDC1 that are produced as a result of alternative splicing events.

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: FNDC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR