Anti-FBXL15 Rabbit Polyclonal Antibody

BS-8475R
10485-174EA 397.94 CAD
10485-174
Anti-FBXL15 Rabbit Polyclonal Antibody
FBXL15 probably recognizes and binds to some phosphorylated proteins and promotes their ubiquitination and degradation.F-box proteins are critical components of the SCF (Skp1-CUL-1-F-box protein) type E3 ubiquitin ligase complex and are involved in substrate recognition and recruitment for ubiquitination. They are members of a larger family of proteins that are involved in the regulation of a wide variety of cellular processes (including the cell cycle, immune responses, signaling cascades and developmental events) through the targeting of proteins, such as cyclins, cyclin-dependent kinase inhibitors. FBL15 (F-box and leucine-rich repeat protein 15), also known as FBXL15, FBXO37, PSD or JET, is a 296 amino acid protein that contains one F-box domain and functions as a component of the SCF complex, possibly playing a role in the ubiquitination and subsequent degradation of target proteins.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: FBXL15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR