Anti-FBXO7 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8489R
10485-676EA 796.88 CAD
10485-676
Anti-FBXO7 Rabbit Polyclonal Antibody
Antibodies
FBXO7, also known as FBX, FBX7 or PKPS, is a 522 amino acid protein that contains one F-box domain and functions as a component of the SCF complex. Defects in the gene encoding FBXO7 are associated with parkinsonian-pyramidal syndrome (PKPS), a hypokinetic rigid disorder that exhibits Parkinsonian and pyramidal-associated symptoms.The FBXO7 gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class and it may play a role in regulation of hematopoiesis. Alternatively spliced transcript variants of this gene have been identified with the full-length natures of only some variants being determined.

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: FBXO7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR