Order Entry
ContactUsLinkComponent
Anti-ELL Rabbit Polyclonal Antibody
Anti-ELL Rabbit Polyclonal Antibody
Catalog #: 10486-240
Supplier:  Bioss
Anti-ELL Rabbit Polyclonal Antibody
Catalog #: 10486-240
Supplier:  Bioss
Price
982,70 $
Log in for availability
Unit of Measure Each (100µl)
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Gene ID:
    8178
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Antigen symbol:
    ELL
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • Size:
    100 µg
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    elongation factor RNA polymerase II,
    Eleven nineteen lysine rich leukemia gene,
    Eleven-nineteen lysine-rich leukemia protein,
    RNA polymerase II elongation factor ELL.,
    RNA polymerase II elongation factor,
    ELL_HUMAN,
    Men,
    ELL1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Cat. no.:
    10486-240

Specifications

About this item

Eukaryotic RNA polymerase II mediates the synthesis of mature and functional messenger RNA. This is a multistep process, called the transcription cycle, that includes five stages: preinitiation, promoter, clearance, elongation and termination. Elongation is thought to be a critical stage for the regulation of gene expression. ELL (11-19 lysine-rich leukemia protein), also designated MEN, functions as an RNA polymerase II elongation factor that increases the rate of transcription by suppressing transient pausing by RNA polymerase II. It is also thought to regulate cellular proliferation. ELL is abundantly expressed in peripheral blood leukocytes, skeletal muscle, placenta and testis, with lower expression in spleen, thymus, heart, brain, lung, kidney, liver and ovary. The gene encoding human ELL, which maps to chromosome 19p13.1, is one of several genes that undergo translocation with the MLL gene on chromosome 11q23 in acute myeloid leukemia. MLL (myeloid/lymphoid leukemia, also designated ALL-1 and HRX) regulates embryonal and hematopoietic development.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ELL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Customers Who Bought This Also Bought