Anti-BARX1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8521R
10485-706EA 806.54 CAD
10485-706
Anti-BARX1 Rabbit Polyclonal Antibody
Antibodies
BarX1 belongs to the Bar subclass of the homeobox gene family. The function of this gene has not yet been determined; however, studies in the mouse and chick homolog suggest a role in developing teeth and craniofacial mesenchyme of neural crest origin. The role of these homologs implicates the human gene as a candidate for unmapped disorders involving tooth and jaw development.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: BARX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR