Anti-CSTF2 Rabbit Polyclonal Antibody

BS-8522R
10486-370EA 430.41 CAD
10486-370
Anti-CSTF2 Rabbit Polyclonal Antibody
Polyadenylation of mRNA precursors is a two-step reaction that requires multiple protein factors. The first step, endonucleolytic cleavage of polyadenylation substrates, requires CstF (cleavage stimulation factor), a heterotrimer that is composed of three distinct subunits. CstF-64 contains an RNA binding domain and is responsible for the RNA binding activity of CstF. CstF-64 is expressed in all somatic cells and in pre- and postmeiotic, but not meiotic, germ cells. However, a large variant of CstF-64, called t CstF-64, is abundantly expressed in meiotic and postmeiotic cells in the testis and to a lesser extent in the brain, and promotes the germ cell pattern of polyadenylation. The gene encoding CstF-64 (designated CSTF2) maps to the X chromosome, whereas t CstF-64 is encoded by an autosomal gene. The increase in CstF-64 concentration during B cell activation switches IgM heavy chain mRNA expression from membrane-bound to secreted forms, suggesting that CstF-64 plays a key role in regulating IgM heavy chain expression during B cell differentiation.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CSTF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR