Anti-TBXAS1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8537R
10662-648EA 796.88 CAD
10662-648
Anti-TBXAS1 Rabbit Polyclonal Antibody
Antibodies
Thymidylate synthase (TS) is an intracellular enzyme critical for de novo synthesis of DNA. In cancer, expression of TS is often elevated and becomes further elevated as a result of treatment with the most commonly used chemotherapeutic, 5-fluorouracil (5-FU). Resistance or lack of response to 5-FU is attributed to the elevation of TS activity. Overexpression of TS has been demonstrated in many types of tumors including breast, colon, gastric and melanoma and is usually predictive of shortened survival time.

  • Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TBXAS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR