Anti-ZNF537 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8540R
10670-664EA 796.88 CAD
10670-664
Anti-ZNF537 Rabbit Polyclonal Antibody
Antibodies
The homeobox DNA-binding domain is a 60 amino acid motif that is conserved among many species and functions to bind DNA via a helix-turn-helix structure, thereby playing a role in transcriptional regulation and the control of gene expression. TSHZ3 (teashirt zinc finger homeobox 3), also known as KIAA1474, TSH3 or ZNF537, is a 1,081 amino acid protein that contains one homeobox DNA-binding domain and five C2H2-type zinc fingers. Localized to the nucleus, TSHZ3 functions as a transcriptional regulator that is involved in developmental processes throughout the body. The gene encoding TSHZ3 maps to human chromosome 19, which consists of over 63 million bases, houses approximately 1,400 genes and is recognized for having the greatest gene density of the human chromosomes.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZNF537
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR