Anti-CTSS Rabbit Polyclonal Antibody

Supplier: Bioss
BS-8558R
10663-030EA 677.18 CAD
10663-030
Anti-CTSS Rabbit Polyclonal Antibody
The cathepsin family of proteolytic enzymes contains several diverse classes of proteases. The cysteine protease class comprises cathepsins B, L, H, K, S, and O (1-6). The aspartyl protease class is composed of cathepsins D and E (7,8). Cathepsin G is in the serine protease class (9). Most cathepsins are lysosomal and each is involved in cellular metabolism, participating in various events such as peptide biosynthesis and protein degradation. Cathepsin S has been shown to be an elastinolytic cysteine proteinase present in aveloar macrophages.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CTSS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR