Anti-RANBP7 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8590R
10663-594EA 796.88 CAD
10663-594
Anti-RANBP7 Rabbit Polyclonal Antibody
Antibodies
Importin 7 is a 1,038 amino acid protein encoded by the human gene IPO7. Importin-7 belongs to the importin b family and contains one importin N-terminal domain. Importin-7 functions in nuclear protein import, either by acting as an autonomous nuclear transport receptor or as an adapter-like protein in association with the Importin b subunit KPNB1. Acting autonomously, Importin-7 is thought to serve itself as receptor for nuclear localization signals (NLS) and to promote translocation of import substrates through the nuclear pore complex (NPC) by an energy requiring, Ran-dependent mechanism. At the nucleoplasmic side of the NPC, Ran binds to Importin-7, the Importin-7/substrate complex dissociates and Importin-7 is re-exported from the nucleus to the cytoplasm where GTP hydrolysis releases Ran. Importin-7 is a nuclear protein that is expressed in most tissues.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: RANBP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR