Anti-IL9 Rabbit Polyclonal Antibody

BS-8591R
10663-618EA 397.94 CAD
10663-618
Anti-IL9 Rabbit Polyclonal Antibody
The protein encoded by this gene is a cytokine that actsas a regulator of a variety of hematopoietic cells. This cytokinestimulates cell proliferation and prevents apoptosis. It functionsthrough the interleukin 9 receptor (IL9R), which activatesdifferent signal transducer and activator (STAT) proteins and thusconnects this cytokine to various biological processes. The geneencoding this cytokine has been identified as a candidate gene forasthma. Genetic studies on a mouse model of asthma demonstratedthat this cytokine is a determining factor in the pathogenesis ofbronchial hyperresponsiveness. [provided by RefSeq, Jul 2008].

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR