Anti-HAO1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8602R
10663-778EA 796.88 CAD
10663-778
Anti-HAO1 Rabbit Polyclonal Antibody
Antibodies
GOX is a 370 amino acid protein that is expressed in liver and pancreas. HAO1 is localized to peroxisomes and aids in organic acid metabolism via 2-hydroxyacid oxidase activity. 2-hydroxyacid oxidases, such as HAO1, are enzymes that require a flavin cofactor to oxidize 2-hydroxyacids to 2-ketoacids while reducing oxygen to hydrogen peroxide. HAO1 prefenentially oxidizes the substrate glycolate and also oxidizes other substrates, including 2-hydroxy fatty acids as well as L-?hydroxy acids of moderately short chain lengths. The oxidation of glycolate yields glyoxylate which is utilized for peroxisomal synthesis of glycine. HAO1 is also able to convert glyoxylate to oxalate. HAO1 is thought to play a role in the pathophysiology of hyperoxaluria type 1, which is caused by defects in AGXT, a peroxisomal enzyme, leading to accumulation of glyoxylate. Hyperoxaluria type 1 is characterized by an accumulation of oxalate that is thought to lead to precipitates of calcium oxalate in kidneys which can be fatal.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: HAO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR