Anti-Visual Arrestin Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8710R
10664-644EA 796.88 CAD
10664-644
Anti-Visual Arrestin Rabbit Polyclonal Antibody
Antibodies
Members of the Arrestin/beta-Arrestin protein family are thought to participate in agonist-mediated desensitization of G protein-coupled receptors, and cause specific dampening of cellular responses to stimuli such as hormones, neurotransmitters or sensory signals. Visual Arrestin, also known as Arrestin, retinal S-antigen or S-Arrestin, is a major soluble photoreceptor protein that regulates light-dependent signal transduction through G protein-coupled receptor (rhodopsin) activation. Visual Arrestin is expressed in retinal photoreceptor cells and the pineal gland. Visual Arrestin is the major pathogenic autoantigen in inflammatory eye disease, such as uveoretinitis and Oguchi disease, a rare autosomal recessive form of night blindness.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: Visual Arrestin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR