Anti-ANKRD6 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8728R
10664-734EA 796.88 CAD
10664-734
Anti-ANKRD6 Rabbit Polyclonal Antibody
Antibodies
Diversin, also known as ANKRD6 (ankyrin repeat domain 6), is a 727 amino acid protein that contains eight ankyrin repeats and belongs to the ankyrin repeat domain protein family. Members of the ankyrin repeat domain family facilitate protein-protein interactions and function as adaptors of signaling pathways. Expressed in a developmentally-regulated manner and at highest levels in the brain, Diversin is believed to play a role in brain development. Via its ankyrin repeats, Diversin can directly interact with Dvl (dishevelled), an interaction that is essential for the activation of noncanonical Wnt signaling. In addition, Diversin contains a C-terminal domain that binds Axin/Conductin and a casein kinase-binding domain in its central region that specifically binds casein kinase Ié. Through the action of these additional domains, Diversin may also facilitate canonical Wnt signaling. Due to alternative splicing events, three Diversin isoforms exist.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ANKRD6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR