Anti-CPOX Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8860R
10664-854EA 796.88 CAD
10664-854
Anti-CPOX Rabbit Polyclonal Antibody
Antibodies
CPOX is a 454 amino acid mitochondrial enzyme that is localized to the inner membrane space of erythrocytes. It participates in the sixth step of heme biosynthesis by catalyzing the formation of protoporphyrinogen IX from copropophyrinogen III. Mutations in the gene encoding CPOX are the cause of coproporphyria, an autosomal dominant disease characterized by skin photosensitivity and neurological disturbances. Symptoms are often experienced as attacks, which include severe abdominal and nerve pain. People affected by coproporphyria overexcrete copropophyrinogen III in feces and urine and the enzymatic activity of CPOX is found to be approximately half that of normal, leading to a decrease in overall heme synthesis. There is no cure for coproporphyria, but preventative treatment to relieve symptoms usually involves dietary changes and avoidance of drugs and alcohol.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CPOX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR