Anti-DUOXA2 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-8863R
10671-308EA 735.06 CAD
10671-308
Anti-DUOXA2 Rabbit Polyclonal Antibody
DUOXA2 is a 320 amino acid multi-pass membrane protein that localizes to the endoplasmic reticulum (ER) and belongs to the DUOXA family. Expressed specifically in thyroid and salivary glands, DUOXA2 is essential for the maturation and transport of DUOX2 from the ER to the plasma membrane and is also thought to play a role in the synthesis of thyroid hormone (TH). Defects in the DUOXA2 gene are associated with the pathogenesis of congenital hypothyroidism, a disorder that affects infants and is characterized by a significant decrease or a complete deficiency of TH from birth. The gene encoding DUOXA2 maps to human chromosome 15, which houses over 700 genes and comprises nearly 3% of the human genome. Angelman syndrome, Prader-Willi syndrome, Tay-Sachs disease and Marfan syndrome are all associated with defects in chromosome 15-localized genes.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: DUOXA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR