Anti-DIS3L2 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9053R
10666-232EA 796.88 CAD
10666-232
Anti-DIS3L2 Rabbit Polyclonal Antibody
Antibodies
The exosome is a multisubunit complex composed of several highly conserved subunits, some of which are 3’ to 5’ exoribonucleases. The complex is involved in a variety of cellular processes and is responsible for degrading unstable mRNAs that contain AU-rich (ARE) elements in their untranslated 3’ region. DIS3L2 (DIS3-like exonuclease 2) is an 885 amino acid protein that is thought to function as an exonuclease and may be required for the 3’ processing of pre-mRNA into mature mRNA. Defects or chromosomal translocations involving the gene encoding DIS3L2 may be associated with Marfanoid habitus, a genetic disorder characterized by abnormalities in the skeleton, eyes and cardiovascular system. DIS3L2 is expressed as five isoforms due to alternative splicing events.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: DIS3L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR