Anti-DXO Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9054R
10666-256EA 796.88 CAD
10666-256
Anti-DXO Rabbit Polyclonal Antibody
Antibodies
DOM3Z, also known as NG6 or DOM3L, is a 396 amino acid ubiquitously expressed protein belonging to the Dom3Z family. The gene encoding DOM3Z maps to human chromosome 6 in the major histocompatibility complex (MHC) class III region. Chromosome 6 contains 170 million base pairs and comprises nearly 6% of the human genome. Deletion of a portion of the q arm of chromosome 6 is associated with early onset intestinal cancer, suggesting the presence of a cancer susceptibility locus. Additionally, Porphyria cutanea tarda, Parkinson's disease and Stickler syndrome are all associated with genes that map to chromosome 6.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: DXO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR